The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028821.3 | NP_083097.2 | 2 - 3 | 112 | 78 |
GenBank mRNA | AK005826.1 | - | 2 - 3 | 198 | 78 |
BC070454.1 | - | - | 69 | 78 | |
BC125392.1 | - | 1 - 2 | 94 | 78 |