The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165256.1 | NP_001158728.1 | 7 - 8 | 834 | 67 |
NM_030246.2 | NP_084522.1 | 8 - 9 | 891 | 67 | |
GenBank mRNA | AK185423.1 | - | 2 - 3 | 100 | 67 |
BC003284.1 | - | 7 - 8 | 800 | 67 | |
CX208460.1 | - | 2 - 3 | 106 | 67 |