Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9
Entrez Gene ID
12279
Gene Name
complement component 9
Gene Aliases
-
Chromosome Location
Chr.15: 6445333 - 6498476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013485.1 NP_038513.1 2 - 3 251 63
GenBank mRNA BC011137.1 - 2 - 3 251 63
X05475.1 - 2 - 3 230 63

View Full Product Details