Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ylpm1
Entrez Gene ID
56531
Gene Name
YLP motif containing 1
Gene Aliases
A930013E17Rik, ZAP, Zap3
Chromosome Location
Chr.12: 84996285 - 85074533 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178363.3 NP_848140.3 12 - 13 5715 119
GenBank mRNA AB033168.1 - 12 - 13 3388 119
AK028731.1 - 2 - 3 150 119
AK044440.1 - 6 - 7 2032 119
AK162250.1 - 4 - 5 424 119

View Full Product Details