Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc34
Entrez Gene ID
216150
Gene Name
cell division cycle 34
Gene Aliases
AI327276, UBE2R1
Chromosome Location
Chr.10: 79682195 - 79688398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK136078.1 - - 171 100

View Full Product Details