The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177355.3 | NP_796329.2 | 2 - 3 | 1035 | 66 |
GenBank mRNA | AK044972.1 | - | 2 - 3 | 882 | 66 |