The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199198.2 | NP_954668.2 | 1 - 2 | 309 | 74 |
NR_028447.1 | 1 - 2 | 309 | 74 | ||
NR_028448.1 | 1 - 2 | 309 | 74 | ||
NR_028449.1 | 1 - 2 | 309 | 74 | ||
GenBank mRNA | AK040235.1 | - | 1 - 2 | 261 | 74 |
AK045028.1 | - | 1 - 2 | 290 | 74 | |
AK154045.1 | - | 1 - 2 | 273 | 74 | |
BC013700.1 | - | 1 - 2 | 262 | 74 | |
BC064018.1 | - | 1 - 2 | 236 | 74 |