Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shank3
Entrez Gene ID
58234
Gene Name
SH3/ankyrin domain gene 3
Gene Aliases
AI841104, SHANK3c-3, SHANK3c-4, Shank3b
Chromosome Location
Chr.15: 89499707 - 89560261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021423.3 NP_067398.2 8 - 9 969 67
GenBank mRNA AB231013.1 - 8 - 9 969 67
AJ245904.1 - 6 - 7 823 67
KF612592.1 - 4 - 5 485 67

View Full Product Details