The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011161.5 | NP_035291.4 | 11 - 12 | 1104 | 99 |
GenBank mRNA | AF135185.1 | - | 11 - 12 | 1084 | 99 |
AK144846.1 | - | 11 - 12 | 1077 | 99 | |
AK161190.1 | - | 9 - 10 | 1279 | 99 | |
AK161562.1 | - | 11 - 12 | 1056 | 99 | |
BC057211.1 | - | 11 - 12 | 1040 | 99 | |
BC064737.1 | - | 11 - 12 | 1055 | 99 | |
BC092526.1 | - | 11 - 12 | 1104 | 99 |