The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027308.2 | NP_081584.1 | 4 - 5 | 947 | 71 |
GenBank mRNA | AK033677.1 | - | 4 - 5 | 948 | 71 |
AK088927.1 | - | 5 - 6 | 903 | 71 | |
AK135657.1 | - | - | 753 | 71 | |
BC139455.1 | - | 4 - 5 | 795 | 71 | |
BC139458.1 | - | 4 - 5 | 795 | 71 |