The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079830.2 | NP_001073299.1 | 16 - 17 | 3514 | 106 |
NM_053170.3 | NP_444400.2 | 16 - 17 | 3514 | 106 | |
GenBank mRNA | AF220138.1 | - | 1 - 2 | 91 | 106 |
AK129293.1 | - | 16 - 17 | 2773 | 106 | |
AY458590.1 | - | 16 - 17 | 3520 | 106 | |
BC043471.1 | - | 3 - 4 | 509 | 106 |