The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301304.1 | NP_001288233.1 | 15 - 16 | 3315 | 118 |
NM_001301306.1 | NP_001288235.1 | 14 - 15 | 3290 | 118 | |
NM_010432.2 | NP_034562.2 | 15 - 16 | 3425 | 118 | |
GenBank mRNA | AF071070.1 | - | 15 - 16 | 3315 | 118 |
AF077658.1 | - | 15 - 16 | 3326 | 118 | |
AF170303.1 | - | 14 - 15 | 3090 | 118 | |
AF170304.1 | - | 15 - 16 | 3225 | 118 | |
AK122333.1 | - | 8 - 9 | 1322 | 118 | |
BC145697.1 | - | 15 - 16 | 3178 | 118 | |
BC145699.1 | - | 15 - 16 | 3178 | 118 |