Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mpped1
Entrez Gene ID
223726
Gene Name
metallophosphoesterase domain containing 1
Gene Aliases
6230403M19, AW456965, D15Bwg0669e
Chromosome Location
Chr.15: 83779472 - 83858487 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172610.3 NP_766198.1 1 - 2 88 107
GenBank mRNA AK048421.1 - 1 - 2 89 107
AK165755.1 - 1 - 2 49 107

View Full Product Details