Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc64b
Entrez Gene ID
212733
Gene Name
coiled-coil domain containing 64B
Gene Aliases
BICDR-2
Chromosome Location
Chr.17: 23660523 - 23668818 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153784.2 NP_722479.1 5 - 6 797 71
GenBank mRNA AK036793.1 - 5 - 6 812 71
AK133729.1 - 5 - 6 814 71
BC038613.1 - 5 - 6 823 71

View Full Product Details