Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc64b
Entrez Gene ID
212733
Gene Name
coiled-coil domain containing 64B
Gene Aliases
BICDR-2
Chromosome Location
Chr.17: 23660523 - 23668818 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153784.2 NP_722479.1 7 - 8 1153 73
GenBank mRNA AK036793.1 - 7 - 8 1168 73
AK133729.1 - 7 - 8 1170 73
BC038613.1 - 7 - 8 1179 73

View Full Product Details