The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008425.1 | NP_001008425.1 | 7 - 8 | 706 | 75 |
GenBank mRNA | BC085131.1 | - | 7 - 8 | 706 | 75 |
BC089551.1 | - | 8 - 9 | 706 | 75 |