Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Flywch1
Entrez Gene ID
224613
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
E030034P13Rik, mKIAA1552
Chromosome Location
Chr.17: 23752797 - 23771591 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153791.2 NP_722486.2 4 - 5 1458 87
GenBank mRNA AK034869.1 - 3 - 4 1052 87
AK052314.1 - 2 - 3 670 87
AK129391.1 - 2 - 3 616 87
AK168389.1 - 4 - 5 1393 87
BC038031.1 - 3 - 4 1750 87

View Full Product Details