Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Flywch1
Entrez Gene ID
224613
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
E030034P13Rik, mKIAA1552
Chromosome Location
Chr.17: 23752797 - 23771591 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153791.2 NP_722486.2 6 - 7 1985 85
GenBank mRNA AK034869.1 - 5 - 6 1579 85
AK052314.1 - 4 - 5 1197 85
AK129391.1 - 4 - 5 1143 85
AK168389.1 - 6 - 7 1920 85
BC025645.1 - 2 - 3 370 85
BC038031.1 - 5 - 6 2277 85

View Full Product Details