Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kremen2
Entrez Gene ID
73016
Gene Name
kringle containing transmembrane protein 2
Gene Aliases
2900054E04Rik, Krm2
Chromosome Location
Chr.17: 23741199 - 23745829 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028416.2 NP_082692.1 9 - 10 1344 68
GenBank mRNA AJ457192.1 - 9 - 10 1356 68
BC120532.1 - 9 - 10 1287 68
BC125438.1 - 9 - 10 1287 68

View Full Product Details