The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023824.3 | NP_076313.1 | 2 - 3 | 691 | 60 |
GenBank mRNA | AK005142.1 | - | 2 - 3 | 714 | 60 |
AK033625.1 | - | 2 - 3 | 709 | 60 | |
AK044092.1 | - | 2 - 3 | 511 | 60 | |
AK044486.1 | - | 2 - 3 | 714 | 60 | |
AK152676.1 | - | 2 - 3 | 690 | 60 | |
AK213384.1 | - | 1 - 2 | 47 | 60 | |
AY424293.1 | - | 2 - 3 | 642 | 60 | |
BC011185.1 | - | 2 - 3 | 691 | 60 | |
BC026921.1 | - | 2 - 3 | 642 | 60 |