Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs6
Entrez Gene ID
50779
Gene Name
regulator of G-protein signaling 6
Gene Aliases
-
Chromosome Location
Chr.12: 82616176 - 83158815 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282061.1 NP_001268990.1 8 - 9 617 96
NM_015812.3 NP_056627.1 8 - 9 899 96
GenBank mRNA AF061933.1 - 5 - 6 344 96
AK052527.1 - 8 - 9 900 96
AK053087.1 - 8 - 9 618 96
AK139323.1 - 8 - 9 1014 96
BC038616.1 - 6 - 7 444 96
BC125029.1 - 8 - 9 603 96
BC125030.1 - 8 - 9 603 96

View Full Product Details