The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020579.2 | NP_065604.2 | 2 - 3 | 255 | 138 |
GenBank mRNA | AF142671.1 | - | 2 - 3 | 195 | 138 |
AK155471.1 | - | 3 - 4 | 355 | 138 | |
BC013619.1 | - | 2 - 3 | 228 | 138 | |
BU703242.1 | - | 2 - 3 | 254 | 138 |