Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb8
Entrez Gene ID
78541
Gene Name
ankyrin repeat and SOCS box-containing 8
Gene Aliases
4930539L19Rik, AI788835, C430011H06Rik
Chromosome Location
Chr.15: 98134637 - 98165602 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170710.1 NP_001164181.1 2 - 3 269 65
NM_001170711.1 NP_001164182.1 2 - 3 275 65
NM_030121.4 NP_084397.3 2 - 3 265 65
GenBank mRNA AF398969.1 - 2 - 3 192 65
AK016001.1 - 2 - 3 323 65
AK021239.1 - 2 - 3 206 65
AK030168.1 - 2 - 3 208 65
BC025106.1 - 2 - 3 189 65
BY240190.1 - 2 - 3 269 65

View Full Product Details