The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025287.2 | NP_079563.2 | 10 - 11 | 1173 | 62 |
GenBank mRNA | AB071989.1 | - | 8 - 9 | 982 | 62 |
AK028201.1 | - | 9 - 10 | 1309 | 62 | |
AK030746.1 | - | 9 - 10 | 1360 | 62 | |
AK132383.1 | - | 9 - 10 | 1349 | 62 | |
AK159482.1 | - | 9 - 10 | 1326 | 62 | |
AK160858.1 | - | 9 - 10 | 1271 | 62 | |
AK166581.1 | - | 9 - 10 | 1283 | 62 | |
AK168185.1 | - | 9 - 10 | 1295 | 62 | |
AK168546.1 | - | 9 - 10 | 1362 | 62 | |
AY538613.1 | - | 8 - 9 | 985 | 62 | |
BC005793.1 | - | 3 - 4 | 292 | 62 | |
BC043131.1 | - | 9 - 10 | 1192 | 62 | |
BC045205.1 | - | 10 - 11 | 1143 | 62 | |
BC064809.1 | - | 9 - 10 | 1297 | 62 |