The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007867.4 | NP_031893.3 | 2 - 3 | 798 | 96 |
GenBank mRNA | AK145123.1 | - | 2 - 3 | 1261 | 96 |
BC106967.2 | - | 2 - 3 | 478 | 96 | |
U73329.1 | - | 2 - 3 | 478 | 96 |