Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Trim25
- Entrez Gene ID
-
217069
- Gene Name
- tripartite motif-containing 25
- Gene Aliases
-
AA960166, AL022677, EFP, Zfp147
- Chromosome Location
-
Chr.11: 88999403 - 89020293 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC006908.1 |
- |
5 - 6 |
|
93 |
View Full Product Details