Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Utp18
Entrez Gene ID
217109
Gene Name
UTP18, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
6230425C22Rik, 6430513M13, Wdr50
Chromosome Location
Chr.11: 93859243 - 93885766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013375.1 NP_001013393.1 12 - 13 1744 130
GenBank mRNA AK032265.1 - 11 - 12 2271 128
AK078033.1 - 7 - 8 880 130
AK154637.1 - 12 - 13 1684 131
BC119621.1 - 12 - 13 1717 129
BC119622.1 - 12 - 13 1717 129

View Full Product Details