Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Utp18
Entrez Gene ID
217109
Gene Name
UTP18, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
6230425C22Rik, 6430513M13, Wdr50
Chromosome Location
Chr.11: 93859243 - 93885766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013375.1 NP_001013393.1 3 - 4 650 140
GenBank mRNA AK032265.1 - 3 - 4 594 140
AK154637.1 - 3 - 4 590 140
AK167248.1 - 3 - 4 649 140
BC119621.1 - 3 - 4 623 140
BC119622.1 - 3 - 4 623 140

View Full Product Details