Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Utp18
Entrez Gene ID
217109
Gene Name
UTP18, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
6230425C22Rik, 6430513M13, Wdr50
Chromosome Location
Chr.11: 93859243 - 93885766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013375.1 NP_001013393.1 6 - 7 932 134
GenBank mRNA AK032265.1 - 6 - 7 876 134
AK078033.1 - 1 - 2 68 134
AK154637.1 - 6 - 7 872 134
AK167248.1 - 6 - 7 931 134
BC119621.1 - 6 - 7 905 134
BC119622.1 - 6 - 7 905 134

View Full Product Details