Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itfg3
Entrez Gene ID
106581
Gene Name
integrin alpha FG-GAP repeat containing 3
Gene Aliases
-
Chromosome Location
Chr.17: 26212692 - 26244242 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207217.4 NP_997100.1 1 - 2 237 87
GenBank mRNA AK029427.1 - 1 - 2 137 87

View Full Product Details