Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Poldip2
Entrez Gene ID
67811
Gene Name
polymerase (DNA-directed), delta interacting protein 2
Gene Aliases
1300003F06Rik, Pdip38
Chromosome Location
Chr.11: 78512296 - 78522736 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026389.3 NP_080665.1 3 - 4 362 76
GenBank mRNA AK165703.1 - 3 - 4 394 76
AK183751.1 - 1 - 2 34 76
BC006833.1 - 3 - 4 362 76
BC012879.1 - 3 - 4 374 76

View Full Product Details