The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172795.3 | NP_766383.2 | 5 - 6 | 1928 | 70 |
GenBank mRNA | AK172964.1 | - | 4 - 5 | 2191 | 70 |
BC052098.1 | - | 5 - 6 | 1574 | 70 | |
BC058534.1 | - | 5 - 6 | 1875 | 70 | |
BC080850.1 | - | 5 - 6 | 1928 | 70 |