Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkl1
Entrez Gene ID
223701
Gene Name
MKL (megakaryoblastic leukemia)/myocardin-like 1
Gene Aliases
AMKL, Bsac, Mal, Mrtf-A
Chromosome Location
Chr.15: 81012281 - 81190757 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082536.1 NP_001076005.1 6 - 7 873 70
NM_153049.3 NP_694629.2 5 - 6 635 70
GenBank mRNA AF385582.1 - 5 - 6 406 70
AF532597.1 - 6 - 7 757 70
AK044188.1 - 6 - 7 866 70
AK089416.1 - 4 - 5 465 70
AK154561.1 - 6 - 7 853 70
AK155821.1 - 6 - 7 876 70
AK155937.1 - 5 - 6 619 70
AK157985.1 - 4 - 5 484 70
BC050941.1 - 6 - 7 766 70
BC054801.1 - 6 - 7 809 70
BY749697.1 - 4 - 5 465 70

View Full Product Details