The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010601.3 | NP_034731.3 | 1 - 2 | 73 | 103 |
GenBank mRNA | AY380579.1 | - | 1 - 2 | 73 | 103 |
BC141013.1 | - | 1 - 2 | 188 | 103 | |
BC145145.1 | - | 1 - 2 | 188 | 103 |