Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpine3
Entrez Gene ID
319433
Gene Name
serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 3
Gene Aliases
E130113E03Rik
Chromosome Location
Chr.14: 62663667 - 62692243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199945.1 NP_001186874.1 2 - 3 324 117
GenBank mRNA AK053602.1 - 2 - 3 326 117

View Full Product Details