Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx7
Entrez Gene ID
76561
Gene Name
sorting nexin 7
Gene Aliases
2510028H01Rik
Chromosome Location
Chr.3: 117781497 - 117868936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029655.3 NP_083931.2 7 - 8 1134 102
GenBank mRNA AK011015.1 - 7 - 8 1109 102
AK038971.1 - 7 - 8 1135 102
AK052487.1 - 7 - 8 1091 102
BC087942.1 - 7 - 8 992 102

View Full Product Details