The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033488.2 | NP_001028660.2 | 5 - 6 | 542 | 58 |
GenBank mRNA | AK133061.1 | - | 5 - 6 | 557 | 58 |
AK156138.1 | - | 6 - 7 | 694 | 58 | |
AK157892.1 | - | 4 - 5 | 1369 | 58 | |
AK162620.1 | - | 5 - 6 | 541 | 58 | |
AK172423.1 | - | 5 - 6 | 543 | 58 |