Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ero1l
Entrez Gene ID
50527
Gene Name
ERO1-like (S. cerevisiae)
Gene Aliases
ERO1-L
Chromosome Location
Chr.14: 45283087 - 45318572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015774.3 NP_056589.1 11 - 12 880 82
GenBank mRNA AK009667.1 - 1 - 2 105 82
AK142595.1 - 11 - 12 871 82
BC025102.1 - 11 - 12 832 82
CT010195.1 - 11 - 12 798 82
CT010259.1 - 11 - 12 798 82

View Full Product Details