Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fermt2
Entrez Gene ID
218952
Gene Name
fermitin family homolog 2 (Drosophila)
Gene Aliases
AA960555, Kindlin-2, Mig2, Plekhc1
Chromosome Location
Chr.14: 45458792 - 45530113 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146054.2 NP_666166.2 4 - 5 669 103
GenBank mRNA AK145030.1 - 4 - 5 611 103
AK165608.1 - 4 - 5 639 103
AK166552.1 - 4 - 5 608 103
BC033436.1 - 4 - 5 590 103
BQ179963.1 - 3 - 4 469 103

View Full Product Details