Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmn2
Entrez Gene ID
54418
Gene Name
formin 2
Gene Aliases
AU024104
Chromosome Location
Chr.1: 174501825 - 174822729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019445.2 NP_062318.2 2 - 3 1937 58
GenBank mRNA AF218940.1 - 2 - 3 1932 58
BC094606.1 - 2 - 3 1937 58

View Full Product Details