Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr64
Entrez Gene ID
75820
Gene Name
WD repeat domain 64
Gene Aliases
4930415O10Rik, 4930511H01Rik
Chromosome Location
Chr.1: 175698569 - 175815734 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029453.2 NP_083729.2 6 - 7 822 85
GenBank mRNA AK133092.1 - 6 - 7 823 85

View Full Product Details