Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fsd2
Entrez Gene ID
244091
Gene Name
fibronectin type III and SPRY domain containing 2
Gene Aliases
9830160G03Rik, Spryd1
Chromosome Location
Chr.7: 81534354 - 81566981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172904.2 NP_766492.2 8 - 9 1489 69
GenBank mRNA AK036674.1 - - 1455 69
AK052866.1 - 2 - 3 289 69

View Full Product Details