The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159706.1 | NP_001153178.1 | 5 - 6 | 753 | 131 |
NM_016770.3 | NP_058050.3 | 5 - 6 | 753 | 131 | |
GenBank mRNA | AK002920.1 | - | 5 - 6 | 720 | 131 |
AK090289.1 | - | - | 754 | 131 | |
BC119604.1 | - | 5 - 6 | 697 | 131 | |
BC119605.1 | - | 5 - 6 | 697 | 131 |