The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081123.1 | NP_001074592.1 | 10 - 11 | 1505 | 97 |
GenBank mRNA | AB073967.1 | - | 10 - 11 | 1372 | 97 |
AK048167.1 | - | 10 - 11 | 1394 | 97 | |
BC090672.1 | - | 10 - 11 | 1476 | 97 | |
BC145645.1 | - | 10 - 11 | 1471 | 97 |