Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cenpf
Entrez Gene ID
108000
Gene Name
centromere protein F
Gene Aliases
6530404A22Rik, AI325968, CENF, Lek1, PRO1779, mitosin
Chromosome Location
Chr.1: 189640606 - 189688105 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081363.2 NP_001074832.2 15 - 16 8273 112
GenBank mRNA AF194970.1 - 4 - 5 6558 112
BC023896.1 - 3 - 4 1025 112
DQ642022.1 - 14 - 15 8130 112

View Full Product Details