Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbt
Entrez Gene ID
13171
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
D3Wsu60e
Chromosome Location
Chr.3: 116513079 - 116549981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010022.3 NP_034152.2 4 - 5 499 102
GenBank mRNA AK075831.1 - 4 - 5 481 102
AK165959.1 - 4 - 5 509 102
BC055890.1 - 4 - 5 455 102

View Full Product Details