Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl29
Entrez Gene ID
208439
Gene Name
kelch-like 29
Gene Aliases
A230106N14Rik, Gm68, Kbtbd9, mKIAA1921
Chromosome Location
Chr.12: 5077465 - 5375682 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164493.1 NP_001157965.1 8 - 9 2277 90
GenBank mRNA AK122572.1 - 8 - 9 1930 90
BC065168.1 - 8 - 9 2277 90
BC138283.1 - 7 - 8 1633 90
BC145230.1 - 7 - 8 1633 90
BC145748.1 - 7 - 8 1633 90

View Full Product Details