Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srsf11
Entrez Gene ID
69207
Gene Name
serine/arginine-rich splicing factor 11
Gene Aliases
0610009J05Rik, 2610019N13Rik, BF642805, Sfrs11
Chromosome Location
Chr.3: 158010493 - 158036650 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001093752.1 NP_001087221.1 9 - 10 1049 80
NM_001093753.1 NP_001087222.1 8 - 9 1031 80
GenBank mRNA AK011472.1 - 9 - 10 987 80
AK081511.1 - 6 - 7 856 80
AK137854.1 - 1 - 2 123 80
AK141191.1 - 2 - 3 161 80
AK146719.1 - 8 - 9 1003 80
BC055763.1 - 10 - 11 936 80
BC069945.1 - 8 - 9 981 80
BC138145.1 - 6 - 7 784 80
BC145162.1 - 6 - 7 784 80

View Full Product Details