The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027861.2 | NP_082137.1 | 1 - 2 | 108 | 60 |
GenBank mRNA | AK050525.1 | - | 1 - 2 | 79 | 60 |
AK146863.1 | - | 1 - 2 | 91 | 60 | |
BB857916.1 | - | 1 - 2 | 109 | 60 | |
BC030340.1 | - | 1 - 2 | 114 | 60 |