The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177380.1 | NP_001170851.1 | 1 - 2 | 173 | 104 |
GenBank mRNA | AK018908.1 | - | 1 - 2 | 173 | 104 |
BC100466.1 | - | 1 - 2 | 173 | 104 |